Antibody & Antigen purification buffers
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J60749.QAEA 50.1 EUR
ALFAJ60749.QA
Antibody & Antigen purification buffers
Physical Form: Liquid
Applications: Buffers for immunoaffinity purification of antibodies.