Ala-OBzl TosOH 98%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L10743.06EA
35.46
EUR
L10743.06
Ala-OBzl TosOH 98%
Ala-OBzl TosOH
Formule:
C₁₇H₂₁NO₅S MW: 351,42 g/mol Smeltpunt: 114…116 °C |
MDL Nummer:
MFCD00066143 CAS nummer: 42854-62-6 EINECS: 255-969-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...