Netherlands
Order Entry
Poly-L-lysine hydrobromide, MP Biomedicals
Poly-L-lysine hydrobromide, MP Biomedicals
Poly-L-lysine hydrobromide, MP Biomedicals
  • CAS No.:
    25988-63-0
  • Storage conditions:
    Freezer
  • Form:
    Powder
  • MDL No.:
    MFCD00237230
  • Protein/peptide name:
    Poly-L-lysine hydrobromide
  • Molecular weight:
    4000 - 20000
  • Molecular formula:
    (C₆H₁₃N₂OBr)n
  •  
    Pack type
    Pk
  • ICNA0210269125
    0210269125
    Amber Glass Bottle
    25 mg
    1 * 25 mg
    1 * 25 mg - 87,70 €
    87,70 €
    1 * 25 mg
       87,70 €
     : 0210269125
     : ICNA0210269125
  • ICNA0210269180
    0210269180
    Amber Glass Bottle
    100 mg
    1 * 100 mg
    1 * 100 mg - 274,00 €
    274,00 €
    1 * 100 mg
       274,00 €
     : 0210269180
     : ICNA0210269180
  • ICNA0210269190
    0210269190
    Amber Glass Bottle
    500 mg
    1 * 500 mg
    1 * 500 mg - 917,00 €
    917,00 €
    1 * 500 mg
       917,00 €
     : 0210269190
     : ICNA0210269190