Quantus® Protect Xtra, Cleanroom Boot Covers
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Quantus®
QNTS083-6PXWH00SIDEA 0 EUR
QNTS083-6PXWH00SID QNTS083-6PXWHM/LID QNTS083-6PXWH00MID QNTS083-6PXWH00LID QNTS083-6PXWH0XLID
Quantus® Protect Xtra, Cleanroom Boot Covers
Werkkleding
These boot covers are made from Protect Xtra fabric which is a further development of the Protect fabric consisting of 100% polyester weave with conductive yarn (warp).
These boot covers are made from Protect Xtra fabric which is a further development of the Protect fabric consisting of 100% polyester weave with conductive yarn (warp). This fabric has been especially designed for use in critical areas (e.g. operating rooms, sterile environments). Thanks to the PTFE finish, the boot covers provide very good splash protection against liquids and other contamination. The fabric has good tensile strength and provides very good resistance to microbial penetration.
  • Plastic zip closure, concealed and offset to the side (outwards)
  • High plastic sole (light grey) is ergonomically moulded, waterproof, autoclavable and provides a roughened profile for skid resistance
  • Reinforcement between sole and fabric, made of CONTROL fabric
  • Gaiter closure at top, adjusted with PES tape and buckle clip
  • Adjustable clip fastener at instep
  • RFID chip included
  • Autoclavable, at approx. 134 °C/ 5 min.
This fabric has been especially designed for use in critical areas (e.g. operating rooms, sterile environments). Thanks to the PTFE finish, the boot covers provide very good splash protection against liquids and other contamination. The fabric has good tensile strength and provides very good resistance to microbial penetration.
  • Plastic zip closure, concealed and offset to the side (outwards)
  • High plastic sole (light grey) is ergonomically moulded, waterproof, autoclavable and provides a roughened profile for skid resistance
  • Reinforcement between sole and fabric, made of CONTROL fabric
  • Gaiter closure at top, adjusted with PES tape and buckle clip
  • Adjustable clip fastener at instep
  • RFID chip included
  • Autoclavable, at approx. 134 °C/ 5 min.
Suitable for use in ISO Class 5 - 8 environments.
Fabric weight: 98,3 g/m²
Bestelinformatie: Other colours and styles are available on request. Please contact your local Avantor office for further information.