Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Cleanroom tape, Ultratape
Leverancier: ULTRATAPE
Polyolefin polypropylene tapes with acrylic adhesive.
  • Ultra low outgassing
  • Low sulfur, low halogen and low heavy metal
  • Adhere lightly to plastics for easy removal
  • Leaves no residue and particulates
Designed specifically for use inside semiconductor and critical environment cleanrooms. It contains extremely low leachable metals, eliminating the concern of contamination.
login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat
Order
Expand All / Collapse All
Omschrijving Lengte Kleur VPE VWR catalogus nummer Hoeveelheid
Polyolefin polypropylene tapes with acrylic adhesive 54,86 m wit 1 Roll UTAP1164WH100P6UU
Polyolefin polypropylene tapes with acrylic adhesive 54,86 m wit 1 Roll UTAP1164WH200P6UU
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid