Avantor® Seradigm, Serum alternative, FB Essence, US origin
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SERD10803-034EA 0 EUR
SERD10803-034 SERD10805-184
Avantor® Seradigm, Serum alternative, FB Essence, US origin
FB Essence is a US origin foetal bovine serum (FBS) alternative that can be used in place of FBS in many cell culture applications. It contains foetal bovine serum, bovine calf serum and equine serum, plus a proprietary blend of supplements including vitamins, minerals and various growth factors.
  • 100% US origin
  • Cost effective alternative to FBS
  • Effective across a broad range of cell types
Avantor Seradigm FBS
Nutritionally superior Sera, proven performance and consistency. Choose your FBS type.
REQUEST YOUR SAMPLE