Manual microplate heat sealer, ALPS™ 30
MATRAB-0400
Specifications
- Description:ALPS™ 30
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:NAR Certificate of Retirement
- Pk:1
Specifications
About this item
Use of the ALPS™ 30 manual heat sealer protects samples from evaporation and contamination when performing PCR or sample storage. Perfect for sealing small numbers of plates in low to medium throughput applications, the ALPS™ 30 unit is small enough to keep on the bench and not take up a lot of space. Its simple, ergonomic manual operation provides quick, efficient sealing.
- Compatible with virtually any height microplate or block, including PCR plates
- Consistent heat seal provides dependable protection for PCR or sample storage plates
- Prevents contamination and evaporation, yielding more reliable results
- Features a built-in thermostat to prevent overheating
- LED indicator lights show when power is on and when unit is heating
- Small footprint so doesn't take up a lot of bench space, and lightweight so easy to carry
- Three adapters included to accommodate most plate types: PCR plates, deep well plates and blocks, standard height 96- or 384-format plates