Netherlands
Order Entry
Fluorescent western blot stripping buffer, Restore™
Fluorescent western blot stripping buffer, Restore™
Certificates
undefined
Fluorescent western blot stripping buffer, Restore™
Product Information
  • Catalogus #
    Leverancier #
    Pk
    Availability
    Price
  • Restore™ Fluorescent Western Blot Stripping Buffer, 20 ml
    Restore™ Fluorescent Western Blot Stripping Buffer, 20 ml
    PIER62299
    62299
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,00 €
    59,00 €
    1 * 1 ST
       59,00 €
    Supplier Article Number: 62299
    Local Article Number: PIER62299
  • Restore™ Fluorescent Western Blot Stripping Buffer, 100 ml
    Restore™ Fluorescent Western Blot Stripping Buffer, 100 ml
    PIER62300
    62300
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 195,00 €
    195,00 €
    1 * 1 ST
       195,00 €
    Supplier Article Number: 62300
    Local Article Number: PIER62300