dNTP set
Leverancier: Omega Bio-Tek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TQAC135EA 161 EUR
TQAC135
dNTP set
Reagentia voor nucleïnezuren Nucleotiden
This dNTP set with dATP, dCTP, dGTP, dTTP is suitable for use in PCR, sequencing, nick translation, fill-in, cDNA synthesis, and TdT tailing reactions, as well as for dilution of radiolabeled dNTPs.