Vacuum manifold, E-Z 96®
Leverancier: Omega Bio-Tek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VAC-03EA 360 EUR
VAC-03
Vacuum manifold, E-Z 96®
Spruitstukken
The E-Z 96® vacuum manifold is designed for DNA/RNA sample preparation with E-Z 96® DNA/RNA plates.
  • Upper plate has a machined recess that fits E-Z 96® filter plate and contains a custom gasket
  • Top plate has locating holes which correspond directly to the locating pins in the bottom plate for easy alignment and stability
The chamber can be used either as a simple dump for waste filtrate or as a receptacle for a standard 96-well microtitre plate (collection plate).