Netherlands
Order Entry
Anti-TCP1 delta Antibody
Anti-TCP1 delta Antibody
  BSBTPB9927
undefined
Anti-TCP1 delta Antibody
  BSBTPB9927
 
552,00 €
   
  1 * 1 EA

 

  • Antibody type:
    Primary
  • Antigen name:
    chaperonin containing TCP1, subunit 4 (delta)
  • Antigen symbol:
    CCT4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    10575
  • Antigen synonyms:
    SRB|Cctd|CCT-DELTA
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for T-complex protein 1 subunit delta(CCT4) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: CCT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat