Donor adult bovine serum, US origin, HyClone™
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HYCLSH30075.03HIEA 197 EUR
HYCLSH30075.03HI HYCLSH30075.03
Donor adult bovine serum, US origin, HyClone™
HyClone™ Donor Adult Bovine Serum, designed to augment cell culture growth, is collected from selected donor herds that receive regular veterinary inspection and care.
  • Cattle are quarantined for 30 days
  • Tested for tuberculosis, brucellosis, and bluetongue virus
  • Serum filtered using sequential 100 nm pore-size rated filters