Netherlands
Order Entry
Buffer solution pH 7, Phosphate buffer
Buffer solution pH 7, Phosphate buffer
Buffer solution pH 7, Phosphate buffer
  • Concentration:
    3.54 g potassium dihydrogen phosphate and 14.7 g i-sodium hydrogen phosphate per litre
  • Appearance:
    Clear colourless solution
  •  
    pH value
    pH accuracy
    pH buffer type
    Pack type
    Pk
  • Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution
    Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution
    ACRO258590025
    258590025
    7,00
    ±0,02
    Phosphate
    Plastic bottle
    2,5 L
    1 * 2,5 L
    1 * 2,5 L - 75,90 €
    75,90 €
    1 * 2,5 L
       75,90 €
  • Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution
    Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution
    ACRO258590010
    258590010
    7,00
    ±0,02
    Phosphate
    Plastic bottle
    1 L
    1 * 1 L
    1 * 1 L - 54,60 €
    54,60 €
    1 * 1 L
       54,60 €
  • Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution
    Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution
    ACRO258591000
    258591000
    7,00
    ±0,02
    Phosphate
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 26,00 €
    26,00 €
    1 * 100 mL
       26,00 €
  • Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution
    Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution
    ACRO258595000
    258595000
    7,00
    ±0,02
    Phosphate
    Plastic bottle
    500 mL
    1 * 500 mL
    1 * 500 mL - 36,70 €
    36,70 €
    1 * 500 mL
       36,70 €