
4-Methylsulphonylbenzylamine hydrochloride 95%
Catalog # ACRO424950025
Supplier: Thermo Fisher Scientific
CAS Number:
Supplier Article Number: 424950025
Local Article Number: ACRO424950025
Price
109,00 €
Log in for availability
Specifications
- Pack type:Glass bottle
- Pk:2,5 g
- Seq:3122
Specifications
Specification Test Results
- Appearance:White to beige to light yellow-pink Powder
- Infrared spectrum:Conforms
- Melting point:269 to 277 °C
- Titration with NaOH:≥94 %
Specification Test Results
About this item
:
:221.70 g/mol
Melting Pt:269…277 °C
:GHS07
Storage Temperature:Ambient
CAS Number:98593-51-2
Formula:C₈H₁₁NO₂S.HCl
MDL Number:MFCD00216491