Netherlands
Order Entry
2,2-Dimethyl-4-pentenal 97% stabilised
Irritant
Flammable
2,2-Dimethyl-4-pentenal 97% stabilised
Catalog # ACRO295040050
CAS Number:  
undefined
Irritant
Flammable
2,2-Dimethyl-4-pentenal 97% stabilised
Catalog # ACRO295040050
CAS Number:  
Supplier Article Number: 295040050
Local Article Number: ACRO295040050
Price
111,00 €
Log in for availability
Unit of Measure 1 * 5 mL

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4200 to 1.4220 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • Stabilizer:
    0.1% Hydroquinone

Specification Test Results

About this item

 :
 :112.172 g/mol
Boiling Pt:124…125 °C
Density:0.825 g/cm³
Flash Pt:18 °C
UN:1989
ADR:3,II
 :GHS07
 :GHS02
Storage Temperature:Refrigerator
CAS Number:5497-67-6
Formula:C₇H₁₂O
MDL Number:MFCD00010699