Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?

(1R,2R)-Bis[(2-methoxyphenyl)phenylphosphino]ethane, 90%

Leverancier: Acros Organics

CAS No.: 55739-58-7

MDL No.: MFCD05863546
GHS Symbols: GHS07
Melting Point: 102°C - 104°C
Appearance: White Crystalline powder
Storage Temperature: Ambient
Signal word: Warning

Formule: C₂₈H₂₈O₂P₂
MW: 458,48 g/mol
Storage Temperature: Kamertemperatuur
MDL Nummer: MFCD05863546
CAS nummer: 55739-58-7
Specification Test Results
Appearance White Crystalline powder

login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat


Expand All / Collapse All
Pack type VPE VWR catalogus nummer Hoeveelheid
Glass bottle 250 mg ACRO379352500
Glass bottle 1 g ACRO379350010
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Smeltpuntapparaat 102°C - 104°C


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid