N-Boc-amino-(4-N-boc-piperidinyl)carboxylic acid 95%

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

ACRO360455000EA 566 EUR
ACRO360455000
N-Boc-amino-(4-N-boc-piperidinyl)carboxylic acid 95%
N-Boc-amino-(4-N-boc-piperidinyl)carboxylic acid

Appearance: White Powder

Formule: C₁₆H₂₈N₂O₆
MW: 344,408 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02683116
CAS nummer: 189321-65-1

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR