Orthophosphoric acid 98+%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7664-38-2
:
Formula:H₃PO₄
MDL Number:MFCD00011340
:98 g/mol
Boiling Pt:∼158 °C (1013 hPa)
Melting Pt:42 °C
Density:1.7 g/cm³
Storage Temperature:Ambient
UN:3453
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Titration with NaOH:≥98.0 %
- Sulfate (SO4):≤30 ppm
- Iron (Fe):≤10 ppm
Specification Test Results
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: 389021000Local Article Number: ACRO389021000
- Supplier Article Number: 389025000Local Article Number: ACRO389025000
- Supplier Article Number: 389020025Local Article Number: ACRO389020025