Storage tubes, barcoded, Matrix™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MATR3815EA 4062 EUR
MATR3815
Storage tubes, barcoded, Matrix™
With permanent 2D barcodes on the base for secure identification and track samples.
  • Tubes sealed using the Thermo Scientific™ SampleSeal™ foil heat sealer
  • Square-welled tube tops are designed for the largest needle or tip access area
  • Leakage proof to ensure sample integrity and security
  • Tracking system includes a 'find' feature to instantly access sample information
  • Compact rack saves space in freezer, refrigerator etc
The proprietary barcoding process produces a permanent, high contrast 2D barcode offering maximum reliability and traceability of compound, biological and genomic samples.