Temperature chart recorder
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Revco®
REVC6383-6EA 2670 EUR
REVC6383-6 REVC6183-5
Temperature chart recorder
Papierbandschrijvers Recorders
Records seven-day history by using pressure sensitive chart paper (150 mm).
  • No inking is required
  • Built-in, panel-mounted recorders are factory installed
  • Free-standing recorders are customer installed