Room climate monitor, RM100
Leverancier: Ebro
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
620-2452EA 174 EUR
620-2452
Room climate monitor, RM100
Bewakingssystemen Bewakingssystemen voor CO2 en temperatuur
For monitoring the CO₂ concentration, the ambient temperature and the relative humidity.
  • Three different LEDs (Green, yellow, red) show the current Indoor Air Quality
  • Helps to improve room climate and prevent mold formation
  • Large LCD with backlight for easy reading in the dark
  • Acoustic alarm when CO₂ limit is exceeded
  • MUTE button for alarm on/off
  • For tabletop use or wall-mounting
This smart, compact and easy-to-use device was designed to help people taking care of the Indoor Air Quality. The RM 100 can be widely used in office buildings, schools, exhibition halls, shopping malls, meeting rooms, fitness centers, restaurants and other public places where personal comfort and a healthy environment is important.
Leveringsinformatie: Supplied with 5V-adapter and Factory calibration certificate.