U heeft gezocht op: Organische standaarden
8 170  results were found
SearchResultCount:"8170"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (42088-100MG)
Leverancier: Merck
Omschrijving: Certified Reference material suitable for HPLC and GC analysis. Produced and certified in accordance with ISO/IEC 17025 and ISO 17034.
UOM: 1 * 100 mg
Catalogus nummer: (C-093-1ML)
Leverancier: Merck
Omschrijving: Organic Standard, Cyclosporin A solution, Ampoule of 1 ml
UOM: 1 * 1 mL
Catalogus nummer: (PHR1371-5ML)
Leverancier: Merck
Omschrijving: Organic Standard, Benzalkonium Chloride 10% Solution
UOM: 1 * 5 mL
Catalogus nummer: (CRM40448)
Leverancier: Merck
Omschrijving: Organic Standard, Limonene solution, Ampoule of 1 ml
UOM: 1 * 1 mL
Catalogus nummer: (PHR1009-1G)
Leverancier: Merck
Omschrijving: Organic Standard, Caffeine
UOM: 1 * 1 g
MSDS
Catalogus nummer: (92264-250MG)
Leverancier: Merck
Omschrijving: Organic Standard, 4-Methoxyphenol, ≥98,5% (GC)
UOM: 1 * 250 mg
Catalogus nummer: (PHR1017-500MG)
Leverancier: Merck
Omschrijving: Organic Standard, Phenylephrine Hydrochloride
UOM: 1 * 500 mg
Catalogus nummer: (90593-10MG)
Leverancier: Merck
Omschrijving: Organic Standard, Demethoxycurcumin, ≥95,0% (HPLC)
UOM: 1 * 10 mg
Catalogus nummer: (CRM40474)
Leverancier: Merck
Omschrijving: Organic Standard, (+)-Menthol solution, Ampoule of 1 ml
UOM: 1 * 1 mL
Catalogus nummer: (D-074-1ML)
Leverancier: Merck
Omschrijving: A stable-labeled internal standard of N-Desmethylcitalopram, an active metabolite of the SSRI antidepressant drug citalopram.
UOM: 1 * 1 mL
Catalogus nummer: (A-094-1ML)
Leverancier: Merck
Omschrijving: Organic Standard, Ascomycin solution, Ampoule of 1 ml
UOM: 1 * 1 mL
Catalogus nummer: (C-116-1ML)
Leverancier: Merck
Omschrijving: Organic Standard, Clomipramine-d3 hydrochloride solution, ampule of 1 ml
UOM: 1 * 1 mL
Catalogus nummer: (C-121-1ML)
Leverancier: Merck
Omschrijving: Organic Standard, Carbamazepine-10,11-epoxide solution, ampule of 1 ml
UOM: 1 * 1 mL
Catalogus nummer: (31737-250MG)
Leverancier: Merck
Omschrijving: Organic Standard, Sulfamethoxazole
UOM: 1 * 250 mg
Catalogus nummer: (CRM47181)
Leverancier: Merck
Omschrijving: Organic Standard, Propionaldehyde-DNPH solution, Ampoule of 1 ml
UOM: 1 * 1 mL
Catalogus nummer: (33497-100MG-R)
Leverancier: Merck
Omschrijving: Organic Standard, Sarafloxacin hydrochloride hydrate
UOM: 1 * 100 mg
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 8 170
no targeter for Bottom