U heeft gezocht op: Cadmium standaarden
26  results were found
SearchResultCount:"26"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: Merck
Omschrijving: Cadmium Standard, 1000 mg/L, Certipur®, Supelco®, Matrix: 0,5 M HNO₃, Toepassingsgebied: AAS standaarden
Leverancier: VWR Chemicals
Omschrijving: Cd in HNO₃ 2 to 5%.
Catalogus nummer: (SUPL36379-100ML-F)
Leverancier: Merck
Omschrijving: Cadmium Standard, 1000 mg/L, TraceCERT®, Supelco®, Matrix: Salpeterzuur verd., Toepassingsgebied: ICP-standaarden
UOM: 1 * 100 mL
Catalogus nummer: (1.70309.0100)
Leverancier: Merck
Omschrijving: Cadmium Standard, 1000 mg/L, Certipur®, Supelco®, Matrix: Salpeterzuur verd., Toepassingsgebied: ICP-standaarden
UOM: 1 * 100 mL
MSDS
Leverancier: VWR Chemicals
Omschrijving: Cadmium Standard, 1000 mg/L, VWR® AVS TITRINORM, Cadmium, Matrix: 2% HNO₃, Toepassingsgebied: AAS standaarden
Leverancier: VWR Chemicals
Omschrijving: Cd in HNO₃ 2 to 5%.
Catalogus nummer: (1.09960.0001)
Leverancier: Merck
Omschrijving: Cadmium Standard, 1000 mg, Titrisol®, Supelco®, Matrix: H₂O, Toepassingsgebied: AAS standaarden
UOM: 1 * 1 Ampul
MSDS
Catalogus nummer: (12313-100ML)
Leverancier: Merck
Omschrijving: Cadmium Standard, 1 mg/L, TraceCERT®, Supelco®, Matrix: Salpeterzuur verd., Toepassingsgebied: ICP-MS-standaarden
UOM: 1 * 100 mL
Catalogus nummer: (5709.0100)
Leverancier: Avantor
Omschrijving: Cadmium Standard, 1000 mg/L, J.T.Baker® BAKER INSTRA-ANALYZED®, J.T. Baker®, Matrix: 5% HNO₃, Toepassingsgebied: ICP-standaarden
UOM: 1 * 100 mL
Leverancier: VWR Chemicals
Omschrijving: Cadmium Standard, 1000 mg/L, VWR® ARISTAR®, Cadmium, Matrix: 2-5% HNO₃, Toepassingsgebied: ICP-MS-standaarden
Leverancier: Thermo Scientific
Omschrijving: Cadmium Standard, 10000 mg/L, Specpure®, Matrix: 5% HNO₃, Toepassingsgebied: ICP-standaarden
Leverancier: Thermo Scientific
Omschrijving: Cadmium Standard, 1000 µg/g, Specpure®, Matrix: Minerale oliën, Toepassingsgebied: Op olie gebaseerde normen
Catalogus nummer: (PERCN9303734)
Leverancier: PerkinElmer
Omschrijving: Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
UOM: 1 * 1 ST
Leverancier: Honeywell Chemicals
Omschrijving: Certified Reference Materials (CRMs)
Catalogus nummer: (HONCE1309-100ML)
Leverancier: Honeywell Chemicals
Omschrijving: Certified Reference Material (CRM)
UOM: 1 * 100 mL
Catalogus nummer: (HONCE1139-100ML)
Leverancier: Honeywell Chemicals
Omschrijving: Certified Reference Materials (CRMs)
UOM: 1 * 100 mL
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 26
no targeter for Bottom