Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
  • Criteria
    • In Stock (1)
  • Leverancier
  • Refine by Suppliers
    Sort by:
  • Zoek binnen resultaten
Erbium standaarden
SearchPresentationType-VERTICAL
SearchResultCount:"12"
  List view Grid view Easy View (new)
Sorteer op:
Leverancier:  Merck
Omschrijving:   Erbium standard solution, 1,000 mg/l Er in dil. nitric acid (from Er2O3) Certipur® standard for ICP
UOM:  1 * 100 ml
Leverancier:  VWR Chemicals
Omschrijving:   [EN]ERBIUM 10 MG/L FOR ICP-MS 1 * 100 ml
Leverancier:  VWR Chemicals
Omschrijving:   Er₂O₃ in HNO₃ 2 - 5%
Leverancier:  VWR Chemicals
Omschrijving:   Erbium standard solution, 1,000 mg/l Er in dil. nitric acid AVS TITRINORM® standard for AAS
Leverancier:  VWR Chemicals
Omschrijving:   Erbium standard solution, 10,000 mg/l Er in dil. nitric acid (from Er2O3) ARISTAR® standard for ICP
UOM:  1 * 500 ml
Leverancier:  Alfa Aesar
Omschrijving:   Erbium standard solution, 10,000 mg/l Er in 5% nitric acid (from Er2O3) Specpure® standard for ICP
Leverancier:  Alfa Aesar
Omschrijving:   Erbium standard solution, 1,000 mg/l Er in 5% nitric acid (from Er2O3) Specpure® standard for ICP
Leverancier:  PERKIN ELMER GC-INSTRUMENTS
Omschrijving:   N/A
UOM:  1 * 125 ml
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 8  Of 12
  1  2  Volgende