Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
  • Criteria
    • In Stock (11)
  • Leverancier
  • Refine by Suppliers
    Sort by:
  • Zoek binnen resultaten
Fosfor standaarden
SearchPresentationType-VERTICAL
SearchResultCount:"21"
  List view Grid view Easy View (new)
Sorteer op:
Leverancier:  Merck
Omschrijving:   Phosphorus standard solution, 1,000 mg/l P in water (from H3PO4) Certipur® standard for ICP
UOM:  1 * 100 ml
Leverancier:  Merck
Omschrijving:   Phosphorus standard solution, 10,000 mg/l P in water (from H3PO4) Certipur® standard for ICP
UOM:  1 * 100 ml
Leverancier:  Merck
Omschrijving:   Standard dissolved in oil is ready-to-use calibration standards for the analysis of phosphorus by AAS in a non-aqueous matrix.
UOM:  1 * 100 ml
Leverancier:  VWR Chemicals
Omschrijving:   NH₄H₂PO₄ in H₂O
Leverancier:  VWR Chemicals
Omschrijving:   NH₄H₂PO₄ in H₂O
Leverancier:  VWR Chemicals
Omschrijving:   Phosphorus standard solution, 1,000 mg/l P in water (from P) ARISTAR® standard for ICP-MS
Leverancier:  VWR Chemicals
Omschrijving:   Phosphorus standard solution, 1,000 mg/l P in water AVS TITRINORM® standard for AAS
Leverancier:  Alfa Aesar
Omschrijving:   Phosphorus standard solution, 1,000 mg/l P in 5% nitric acid (from P) Specpure® standard for ICP
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 8  Of 21
  1  2  3  Volgende