Netherlands
Order Entry
Microplate handling system, StakMax®
Microplate handling system, StakMax®
Supplier:  Molecular Devices
Certificates
undefined
Microplate handling system, StakMax®
Supplier:  Molecular Devices
Specifications
  • Catalog No:
  • MLDVSTAKMAX
  • MLDV0200-6016
  • MLDV0200-6017
  • MLDV0200-6187
  • MLDV5005331
  • Model:
  • StakMax®
  • Interfaces:
  • USB 2.0
  • Plate types:
  • 96- to 384-well plates
  • Temperature operating range:
  • 10 - 40 °C
  • W×D×H:
  • 440×509×231 mm
  • Weight:
  • 11,8 kg
Specifications
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • StakMax® microplate handling system
    StakMax® microplate handling system
    MLDVSTAKMAX
    STAKMAX
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Model
    StakMax®
    Environmentally Preferable
    Interfaces
    USB 2.0
    Plate types
    96- to 384-well plates
    Temperature operating range
    10 - 40 °C
    W×D×H
    440×509×231 mm
    Weight
    11,8 kg
Accessories
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • 20-plate stacker magazines (pair)
    20-plate stacker magazines (pair)
    MLDV0200-6016
    0200-6016
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    Specifications
    Model
    Environmentally Preferable
    Interfaces
    Plate types
    Temperature operating range
    W×D×H
    Weight
  • 40-plate stacker magazines (pair)
    40-plate stacker magazines (pair)
    MLDV0200-6017
    0200-6017
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    Specifications
    Model
    Environmentally Preferable
    Interfaces
    Plate types
    Temperature operating range
    W×D×H
    Weight
  • 50-plate stacker magazines (pair)
    50-plate stacker magazines (pair)
    MLDV0200-6187
    0200-6187
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    Specifications
    Model
    Environmentally Preferable
    Interfaces
    Plate types
    Temperature operating range
    W×D×H
    Weight
  • Loading tool for facilitating loading and unloading plates
    Loading tool for facilitating loading and unloading plates
    MLDV5005331
    5005331
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 230,00 €
    2 230,00 €
    1 * 1 ST
       2 230,00 €
    Specifications:
    Specifications
    Model
    Environmentally Preferable
    Interfaces
    Plate types
    Temperature operating range
    W×D×H
    Weight