Microplate readers, SpectraMax® 190 absorbance plate reader
Leverancier: Molecular Devices
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MLDV190EA 25979 EUR
MLDV190
Microplate readers, SpectraMax® 190 absorbance plate reader
SpectraMax® 190 absorbance plate reader enables data acquisition in endpoint, kinetic, and spectral scan modes at wavelengths across the UV-visible wavelength range, with the only temperature-independent method of pathlength correction. Temperature control and shaking come as standard.
  • Read up to 6 wavelengths in endpoint or kinetic mode for 96-well plates, and perform spectral scans to determine wavelength peaks
  • Measurement accuracy is increased due to narrow bandwidths that deliver increased and linearity over the widest range of assays, including DNA quantitation in the UV range
  • Thorough mixing of samples in microplate wells by user-programmable Automix feature
  • Quickly quantify DNA, RNA, and protein samples as low as 2 µl with the SpectraDrop® micro-volume microplate
The patented multichannel optical design of the SpectraMax® 190 absorbance microplate reader mimics a dual beam spectrophotometer, measures each sample in the plate directly, and eliminates measurement error due to variations in light output between optical fibers on other instruments for more reliable data reporting. The eight-channel system delivers both high precision and speed when reading 96-well microplates for endpoint measurements, kinetic assays, and spectral scans. Narrow, 2 nm bandwidths deliver increased measurement accuracy and linearity over the widest range of assays, including ELISAs in the visible range or DNA quantitation in the UV range.
Leveringsinformatie: Includes current version of SoftMax® Pro software. Additional accessories are available including the µMax Low Volume Plate, standard validation tools, and 21 CFR Part 11 Compliance tools, please contact your local Avantor sales office for more details.