Potassium bromide die set
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PERC51001144EA 978 EUR
PERC51001144
Potassium bromide die set
Persmachines
The evacuable potassium bromide (KBr) die is designed to produce uniform KBr pellets.
Suitable for FT-IR spectrometry of solid samples.