CaptairFlex® XLS, ductless fume hoods
Leverancier: Erlab
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CaptairFlex® XLS
ERLAI2820000003EA 395 EUR
ERLAI2820000003
CaptairFlex® XLS, ductless fume hoods
Mobile ductless filtration fume hood for personal protection during handlings of liquid, gaseous and solid chemicals. Molecular filtration by adsorption on activated carbon filter and particulate filtration on H14 HEPA filter.
  • Automatic filter saturation alarm
  • Security carbon filter allowing maximum security
  • Permanent air flow and air face velocity monitoring system
  • Adjustable timer for periodical saturation tests
  • Programmable timer