Freeze dryer, benchtop, LYO GT2
Leverancier: SRK Systemtechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
535-2114EA 0 EUR
535-2109 535-2107 535-2108 535-2105 535-2106 535-2114 535-2103 535-2213 535-2104 535-2211 535-2112 535-2101 535-2212 535-2102 535-2113 535-2110 535-2111
Freeze dryer, benchtop, LYO GT2
Vriesdrogers
LYO GT2 Laboratory freeze-dryers are usually used in the areas of pharmacy, biotechnology and medicine.
  • Used for the conservation of temperature-sensitive products
  • Suitable for the freeze-drying of products in flasks, ampoules, vials or trays
  • Made of corrosion-resistant stainless steel
  • Equipped with a microprocessor-controlled operating module with a large LCD display
The two-chamber design permits carrying out pressure rise measurements (PRM) in the drying chamber to determine the end of the drying process. This design also permits carrying out barometric temperature measurements (BTM).
The basic unit of the device includes an air cooled compressor, stainless steel condenser and a microprocessor-based system controller that fully controls the processor to create a user-friendly whole.
LYOVAC GT2 is equipped with a modern microprocessor control with clear, easy-to-use control panel. The controller has RS232 interface for PC connection, output to recorder, control of heated shelves, vacuum pump and pressure regulator (vacuum).