Cleanroom face masks, Sierra™, 7360
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DuPont™ Sierra™
DUPOML7360WHBHEA 207 EUR
DUPOML7360WHBH
Cleanroom face masks, Sierra™, 7360
Maskers Cleanroom Maskers
This face mask combines high breathability with exceptional filtration efficiency.
  • Bound ties for cleanroom use
  • Pleated rayon outer facing
  • Metal nose piece
  • Gamma sterilised to SAL of 10⁻⁶ (only sterile version)
  • Bacterial Filtration Efficiency (BFE): 98,4 - 99,7 (ASTM F2101)
  • Particle Filtration Efficiency (PFE): 98,4 - 99,5 (ASTM F2299)
Suitable for use in Class 100, ISO Class 5, M3.5 cleanrooms.
Verpakking: Sterile mask is individually packed (250 per case). Non sterile mask is bulk packed (5 bags of 50).