Cleanroom shoe covers, Tyvek® IsoClean®, model IC 451 S
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DUPOIC451SWH0B/MEA 169 EUR
DUPOIC451SWH0B/M
Cleanroom shoe covers, Tyvek® IsoClean®, model IC 451 S
These shoe covers are made of Tyvek®. This fabric is composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
  • Ultra-low-linting fabric
  • With elastic top
  • Gripper™ soles provide enhanced skid-resistance and durability
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size). Ideal for use in biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other critical or controlled environments.
Certificaten: Type PB [6-B]; EN 14126; Cat. III; (EU) 2016/425
Verpakking: Bulk packaged in double transparent poly liners.