Micrometer film applicators
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIMELD3571EA 986 EUR
SIMELD3571
Micrometer film applicators
These adjustable micrometer film applicators are ergonomically designed to facilitate fast, accurate draw downs. Ideally suited to research projects where small variations in film thickness need to be evaluated accurately.
  • No springs to break, rust or get clogged
  • Detachable blades for easy cleaning
  • Replacement blades are easy to install and calibrate
  • Enhanced accuracy
Applicators have a finely machined stainless steel knife blade that fits into slots in the end section, allowing vertical adjustment of the blade. The two micrometer barrels can be easily set so that gate clearances can be adjusted up to 6,35 mm (0,01 mm per graduation, accuracy ±0,005 mm). Thimbles and barrels of the micrometers are dull chrome.
The clear anodised aluminium frame has two end sections that provide a large paint well. The end sections also serve as ergonomic operator hand grips when drawing down wet films. Rare earth magnets hold the stainless steel blade firmly in place allowing for quick, easy detachment and cleaning.