Avantor® Seradigm Bovine Calf Serum (BCS), Iron-Supplemented, US Origin
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Avantor® Seradigm
10158-358EA 0 EUR
10158-358 10158-360
Avantor® Seradigm Bovine Calf Serum (BCS), Iron-Supplemented, US Origin
Serum
Iron-supplemented bovine calf serum, sourced from US origin animals aged up to six months, and raised on a diet consisting mainly of formula.
  • 100% US origin
  • Cost effective alternative to FBS
  • Similar performance to FBS in many established cell lines
These source animals have naturally high levels of transferrin. When the serum is supplemented with iron, the iron binds to the unbound transferrin, resulting in a product with three to four times the amount of available iron compared to FBS or equine serum. This product is an excellent, cost effective alternative to FBS and often performs as well as FBS in many established cell lines. In some applications, it even outperforms FBS and equine serum.
Certificaten: International Serum Industry Association (ISIA) certified supply chain for traceability.
Avantor Seradigm FBS
Nutritionally superior Sera, proven performance and consistency. Choose your FBS type.
REQUEST YOUR SAMPLE