Distance gauges, Leica DISTO™
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIMEDM2636EA 481 EUR
SIMEDM2636
Distance gauges, Leica DISTO™
The Leica DISTO™ provide a combination of distance meter and tilt sensor in indirect measurements of distance and heights.
  • Large colour display
  • Long distances measurements with digital zoom (4×)
  • Dimension marked using arrows in the display