Low voltage pinhole detectors
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIMELD8105EA 531 EUR
SIMELD8105 SIMELD8100
Low voltage pinhole detectors
These pinhole detectors can inspect coatings on conductive substrates for small defects.
  • Visual, audible and tactile feedback
  • Return cable connectivity detection
  • Smart power saving features ie. screen dimming, stand-by and automatic power off
  • Detected pinhole counter with automated self diagnostics
Certificaten: ISO 8289-A, 14654:1999 ; BS 7793-2:1996 ; ASTM D 5162-A ; JIS K 6766:2008 ; TM0384-2002
Leveringsinformatie: Supplied with sponge wand, grounding cable with clamp, calibration certificate and user manual.