Avantor® ACE® NH₂, HPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
These stainless steel columns with proprietary bonding technology provide high robustness with excellent column lifetimes.
- Usable in reversed phase, normal phase, weak anion-exchange and HILIC modes
- Ideal for the analysis of sugars
- Ultra-inert, high efficiency with optimum peak shape
- Range of particle sizes: 1,7 µm, 3 µm and 5 µm and complementary phases
Specifications
- Catalog No:
- ACE-1214-2530
- ACE-1214-1220
- ACE-1214-0510
- ACE-1214-3030
- ACE-1214-0520
- ACE-1214-2510
- ACE-1214-7530
- ACE-1214-2520
- ACE-1214-1530
- ACE-1214-7520
- ACE-1214-1030
- ACE-1214-1510
- ACE-1214-1520
- ACE-1214-2508
- ACE-1214-1020
- ACE-1214-0530
- ACE-1214-1010
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- 4,0%
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Max. pressure:
- 350 bar
- 350 bar
- 350 bar
- 350 bar
- 350 bar
- 350 bar
- pH range:
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- USP code:
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Product Information
- Catalog #Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
- 1/16" connections1/16" connectionsACE-1214-2510Semi-preparativeNH₂100 Å5 µm10,00 mm250 mmLog in for availability1 * 1 STLocal Article Number: ACETACE-1214-2510Specifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load4,0%EndcappedNoMax. pressure350 barpH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 1/16" connections1/16" connectionsACE-1214-1520PreparativeNH₂100 Å5 µm21,20 mm150 mmLog in for availability1 * 1 ST4 650,00 €Local Article Number: ACETACE-1214-1520Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load4,0%EndcappedNoMax. pressure350 barpH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certification - 1/16" connectionsLocal Article Number: ACETACE-1214-2520Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load4,0%EndcappedNoMax. pressure350 barpH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certification - 1/16" connectionsLocal Article Number: ACETACE-1214-2530Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load4,0%EndcappedNoMax. pressure350 barpH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certification - 1/16" connectionsLocal Article Number: ACETACE-1214-3030Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load4,0%EndcappedNoMax. pressure350 barpH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certification - 1/16" connections1/16" connectionsACE-1214-2508Semi-preparativeNH₂100 Å5 µm7,75 mm250 mmLog in for availability1 * 1 ST1 560,00 €Local Article Number: ACETACE-1214-2508Specifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load4,0%EndcappedNoMax. pressure350 barpH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Local Article Number: ACETACE-1214-1220Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load4,0%EndcappedNoMax. pressurepH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certification - Local Article Number: ACETACE-1214-0510Specifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load4,0%EndcappedNoMax. pressurepH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Local Article Number: ACETACE-1214-0520Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load4,0%EndcappedNoMax. pressurepH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certification - Local Article Number: ACETACE-1214-7530Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load4,0%EndcappedNoMax. pressurepH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certification - Local Article Number: ACETACE-1214-1530Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load4,0%EndcappedNoMax. pressurepH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certification - Local Article Number: ACETACE-1214-7520Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load4,0%EndcappedNoMax. pressurepH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certification - Local Article Number: ACETACE-1214-1030Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load4,0%EndcappedNoMax. pressurepH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certification - Local Article Number: ACETACE-1214-1510Specifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load4,0%EndcappedNoMax. pressurepH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Local Article Number: ACETACE-1214-1020Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load4,0%EndcappedNoMax. pressurepH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certification - Local Article Number: ACETACE-1214-0530Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load4,0%EndcappedNoMax. pressurepH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certification - Local Article Number: ACETACE-1214-1010Specifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load4,0%EndcappedNoMax. pressurepH range2,0 - 7,0Surface area300 m²/gUSP codeL8Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Guard holders and accessories
Product Information
- Catalog #DescriptionPkAvailabilityPrice
- Local Article Number: ACETC0002