Polyethyleneimine, M.W. 50,000 - 100,000 50% in aqueous solution, viscous solution
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9002-98-6
:
Formula:(CH2CH2NH)n
MDL Number:MFCD00084427
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- Concentration:~50% w/v
Specification Test Results
Product Information
- Catalog #Supplier #PkAvailabilityPrice