Cleaning trolley Clino®, CR4 EM-GMP
Leverancier: Pfennig
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
141-1209EA 2658.8 EUR
141-1209
Cleaning trolley Clino®, CR4 EM-GMP
Transportwagens
The Clino® CR4 EM-GMP trolley system made of stainless steel suitable for cleanrooms and for high sensitive areas that require minimal space.
  • Individually configurable through Clino modular design system
  • EasyMop GMP with system box certified for areas under sterille conditions
  • Highest efficiency and process reliability in single step technique
  • Including contact-free dropping device MopDrop
  • Basic module mini with high end finish and castors (100 mm)
The trolley is an high-end solution for the cleanroom cleaning in low classified cleanrooms up to sterile areas. The EasyMop pre-preparation systems allows a validatable and reproducible cleaning and disinfection of surfaces.
Certificaten: With IPA certificate.
Leveringsinformatie: Supplied with 2×system boxes EasyMop GMP, 2×4-litre stainless steel container, disposal unit (1×35 L) and stainless steel container (9 L).