Gasdistributiebuisjes
Leverancier: Lenz
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LENZ07212025EA 40.5 EUR
LENZ07212025 LENZ07212125 LENZ07212225
Gasdistributiebuisjes
Gasbuisjes
Gas distribution tubes with central ring made of borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion