About this item
Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raise the possibility of paraganglioma. The co-expression of chromogranin and NSE is typical of neuroendocrine neoplasms. Most pituitary adenomas and prolactinomas readily express chromogranin.
Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: PHE5
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey
Specifications
- Catalog No:
- BNC940461-500
- BNC940461-100
- Antigen symbol:
- CHGA
- CHGA
- Antigen name:
- Chromogranin A
- Chromogranin A
- Antigen synonyms:
- Pituitary Secretory Protein I|Chromogranin A Parathyroid Secretory Protein 1|ER-37|Vasostatin I or II|Parastatin|SP-I|Pancreastatin|CHGA|Beta-Granin|CGA
- Pituitary Secretory Protein I|Chromogranin A Parathyroid Secretory Protein 1|ER-37|Vasostatin I or II|Parastatin|SP-I|Pancreastatin|CHGA|Beta-Granin|CGA
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 594
- CF® 594
- Clone:
- Clone PHE5
- Clone PHE5
- Reactivity:
- Monkey, Human
- Monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 150793
- 150793
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Product Information
- Catalog #Supplier #SizePkAvailabilityPrice
- Supplier Article Number: BNC940461-500Local Article Number: BTIUBNC940461-500Specifications:
More
Specifications
Antigen symbolCHGAAntigen nameChromogranin AEnvironmentally PreferableAntigen synonymsPituitary Secretory Protein I|Chromogranin A Parathyroid Secretory Protein 1|ER-37|Vasostatin I or II|Parastatin|SP-I|Pancreastatin|CHGA|Beta-Granin|CGAAntibody typePrimaryClonalityMonoclonalConjugationCF® 594CloneClone PHE5ReactivityMonkey, HumanHostMouseGene ID150793IsotypeIgG1 kappaImmunoChemistryYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide - Supplier Article Number: BNC940461-100Local Article Number: BTIUBNC940461-100Specifications:
More
Specifications
Antigen symbolCHGAAntigen nameChromogranin AEnvironmentally PreferableAntigen synonymsPituitary Secretory Protein I|Chromogranin A Parathyroid Secretory Protein 1|ER-37|Vasostatin I or II|Parastatin|SP-I|Pancreastatin|CHGA|Beta-Granin|CGAAntibody typePrimaryClonalityMonoclonalConjugationCF® 594CloneClone PHE5ReactivityMonkey, HumanHostMouseGene ID150793IsotypeIgG1 kappaImmunoChemistryYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide