Netherlands
Order Entry
Anti-CNN1 Mouse Monoclonal Antibody (CF594) [clone: CNN1/832]
Anti-CNN1 Mouse Monoclonal Antibody (CF594) [clone: CNN1/832]
Supplier:  Biotium
Certificates
Anti-CNN1 Mouse Monoclonal Antibody (CF594) [clone: CNN1/832]
Supplier:  Biotium
Specifications
  • Catalog No:
  • BNC940832-500
  • BNC940832-100
  • Antigen symbol:
  • CNN1
  • CNN1
  • Antigen name:
  • Calponin-1
  • Calponin-1
  • Antigen synonyms:
  • Calponin-1|Calponin 1 basic smooth muscle|Sm Calp|CNN1|SMCC|Cnn1|Calponin H1 smooth muscle
  • Calponin-1|Calponin 1 basic smooth muscle|Sm Calp|CNN1|SMCC|Cnn1|Calponin H1 smooth muscle
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone CNN1/832
  • Clone CNN1/832
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 465929
  • 465929
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide
Specifications
Product Information
  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BNC940832-500
    BNC940832-500
    500 μL
    500 µl
    Log in for availability
    Log in for availability
    1 * 500 µl
    1 * 500 µl - 560,00 €
    560,00 €
    1 * 500 µl
       560,00 €
    Supplier Article Number: BNC940832-500
    Local Article Number: BTIUBNC940832-500
    Specifications:
    More
    Specifications
    Antigen symbol
    CNN1
    Antigen name
    Calponin-1
    Environmentally Preferable
    Antigen synonyms
    Calponin-1|Calponin 1 basic smooth muscle|Sm Calp|CNN1|SMCC|Cnn1|Calponin H1 smooth muscle
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 594
    Clone
    Clone CNN1/832
    Reactivity
    Human, Rat
    Host
    Mouse
    Gene ID
    465929
    Isotype
    IgG1 kappa
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNC940832-100
    BNC940832-100
    100 μL
    100 µl
    Log in for availability
    Log in for availability
    1 * 100 µl
    1 * 100 µl - 203,00 €
    203,00 €
    1 * 100 µl
       203,00 €
    Supplier Article Number: BNC940832-100
    Local Article Number: BTIUBNC940832-100
    Specifications:
    More
    Specifications
    Antigen symbol
    CNN1
    Antigen name
    Calponin-1
    Environmentally Preferable
    Antigen synonyms
    Calponin-1|Calponin 1 basic smooth muscle|Sm Calp|CNN1|SMCC|Cnn1|Calponin H1 smooth muscle
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 594
    Clone
    Clone CNN1/832
    Reactivity
    Human, Rat
    Host
    Mouse
    Gene ID
    465929
    Isotype
    IgG1 kappa
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide