Anti-PPCDC Rabbit Polyclonal Antibody
Leverancier: Abgent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABGEAP13975AEA 348 EUR
ABGEAP13975A
Anti-PPCDC Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: PPCDC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse