Storage vessel for electrodes
Leverancier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
662-1155EA 45.3 EUR
662-1155
Storage vessel for electrodes
Elektrodehouders
Plastic receptacle with squeezing ring sealing and bayonet fastener for electrodes with 12 mm diameter.