Drum truck, universal
Leverancier: New Pig
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NEWPDRME200EA 0 EUR
NEWPDRME200
Drum truck, universal
Ideal for multiple drum sizes and types like steel, plastic, fibre and salvage drums. Retaining strap with tensioner and safety catch holds drums securely against truck.
  • Blue powder coated finish adds durability and helps prevent rusting
  • Steel footplate slides under base of drum, easily lifting even full drums when truck is tipped back
  • Large, rubber cushion wheels with roller bearings offer smooth travel in a variety of conditions
  • Load capacity of 250 kg