Glass tubings
Leverancier: Hecht
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HECH42840001EA 9.8 EUR
HECH42840001
Glass tubings
AR-Glass®, clear.
  • Good chemical and physical properties
  • Suitable for the production of items that must normally withstand only brief exposure to aggressive chemicals
  • Not resistant to high thermal loads
Tube length ~1500 mm