96-well plates, glass, for use with LPO assay kit
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CAYM400015-1EA 1300 EUR
CAYM400015-1
96-well plates, glass, for use with LPO assay kit
Microplaten Platen voor vastefase-extractie
These 96-well glass plates are for use with the Lipid Hydroperoxide (LPO) assay kit. To prepare for use, clean the plate with warm soapy water and then rinse it with HPLC-grade water, followed by rinsing with acetone.
  • Reusable, must be cleaned before using each time
  • For laboratory research use only
  • Not for human or veteinary use