U heeft gezocht op: Containers voor scherpe materialen
14  results were found
SearchResultCount:"14"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: Hospidex
Omschrijving: Sharpsafe®, historic pioneer in protecting against the risk of sharps injuries and sustainability in medical waste packaging management, presents the ...
Afvalvermindering
Duurzame materialen
Energiezuinig
Lage impact op de fabricage
Producttransparantie en openbaarmaking
Veiligere gezondheid van mens en milieu
Catalogus nummer: (HUARSHA-41912430)
Leverancier: Hospidex
Omschrijving: The Clinisafe® plastic clinical waste containers have been designed to hold clinical, anatomical and laboratory waste.
UOM: 1 * 20 ST
Leverancier: Hospidex
Omschrijving: Made from 30% recycled plastic. Fully opening lid, with original positioning of hinge.
Leverancier: Synergy Health
Omschrijving: All SHARPAKâ„¢ Code YELLOW sharps containers feature yellow coloured closures and are recommended for the segregation, collection and disposal of partia...
Leverancier: Synergy Health
Omschrijving: All SHARPAKâ„¢ Code ORANGE sharps containers feature orange coloured closures and are recommended for the segregation, collection and disposal of used a...
Catalogus nummer: (119-4264)
Leverancier: DELTALAB
Omschrijving: Needle collection containers are made of high density polyethylene suitable for waste type III.
UOM: 1 * 1 ST
Leverancier: B. Braun
Omschrijving: Medibox® containers are suitable for easy and safe disposal of all kinds of medical sharps. Consist of diverse ports for disposal without contact. Com...
Catalogus nummer: (GREI450332_30)
Leverancier: Greiner Bio-One
Omschrijving: Disposal containers, PP, yellow, non-sterile, are safe disposal means for needles, sharps and dangerous objects.
UOM: 1 * 30 ST
Leverancier: HEATHROW
Omschrijving: This sharps container provides safe disposal of syringes, needles and other small sharps. Easy to use and transport.
Catalogus nummer: (LPIT138013)
Leverancier: LP ITALIANA SPA
Omschrijving: Sharp disposal container, plastic, with special lid for the extraction of needles.
UOM: 1 * 40 ST
Catalogus nummer: (DELT232806)
Leverancier: DELTALAB
Omschrijving: Manufactured in PP, these containers are also shockproof, perforation and solvents resistant.
UOM: 1 * 100 ST
Leverancier: SARSTEDT
Omschrijving: PP, yellow/red.
Catalogus nummer: (SARS77.3890.002)
Leverancier: SARSTEDT
Omschrijving: Boxes, made of PP, yellow/red. Secure sharp container with final closure in a handy size for doctors, nurses, rescue services- and ambulatory medical ...
UOM: 1 * 110 ST
Market Source Item This is a MarketSource item. Additional charges may apply
Catalogus nummer: (SARS77.3899.060)
Leverancier: SARSTEDT
Omschrijving: Box, made of PP, yellow/red. Secure and robust container for the disposal of infectious wastes that can be deactivated by autoclaving at 121 or 134 °C...
UOM: 1 * 120 ST
Market Source Item This is a MarketSource item. Additional charges may apply
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 14 of 14
no targeter for Bottom