Netherlands
Order Entry
Screw caps, DURAN® PURE Premium
Screw caps, DURAN® PURE Premium
Supplier:  DWK Life Sciences
Certificates
undefined
Screw caps, DURAN® PURE Premium
Supplier:  DWK Life Sciences
Certificates
Product Information
  • Catalog #
    Supplier #
    Thread
    Pk
    Availability
    Price
  • DURAN® PURE Premium screw caps, TpCh260 TZ, with PTFE coated silicone seal (temperature resistant from−196…+200 °C)
    DURAN® PURE Premium screw caps, TpCh260 TZ, with PTFE coated silicone seal (temperature resistant from−196…+200 °C)
    215-3739
    891081306
    GL 25
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 40,40 €
    40,40 €
    1 * 5 ST
       40,40 €
    Supplier Article Number: 891081306
    Local Article Number: SCOT891081306
    Specifications:
    Specifications
    Environmentally Preferable
  • DURAN® PURE Premium screw caps, TpCh260 TZ, with PTFE coated silicone seal (temperature resistant from −196…+200 °C)
    DURAN® PURE Premium screw caps, TpCh260 TZ, with PTFE coated silicone seal (temperature resistant from −196…+200 °C)
    215-3740
    891088679
    GL 45
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,00 €
    16,00 €
    1 * 1 ST
       16,00 €
    Supplier Article Number: 891088679
    Local Article Number: SCOT891088679
    Specifications:
    Specifications
    Environmentally Preferable
  • DURAN® PURE Premium screw caps, PFA, with lip seal (temperature resistant −196…+200 °C)
    DURAN® PURE Premium screw caps, PFA, with lip seal (temperature resistant −196…+200 °C)
    215-4773
    898862802
    GL 45
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 99,10 €
    99,10 €
    1 * 5 ST
       99,10 €
    Supplier Article Number: 898862802
    Local Article Number: SCOT898862802
    Specifications:
    Specifications
    Environmentally Preferable
  • DURAN® PURE pouring ring, GL 45, TpCh260 TZ (temperature resistant −196…+200 °C)
    DURAN® PURE pouring ring, GL 45, TpCh260 TZ (temperature resistant −196…+200 °C)
    SCOT891088678
    891088678
    GL 45
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 21,10 €
    21,10 €
    1 * 5 ST
       21,10 €
    Supplier Article Number: 891088678
    Local Article Number: SCOT891088678
    Specifications:
    Specifications
    Environmentally Preferable