Hot swap capillary column nuts
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
REST22348EA 89 EUR
REST22348
Hot swap capillary column nuts
Bevestigingen Moeren
This hot swap capillary column nut allows to change capillary column while the injector temp is still hot without downtime waiting for injector parts to cool down.